Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa2
Entrez Gene ID
498528
Gene Name
shisa family member 2
Gene Aliases
RGD1561256, Tmem46
Chromosome Location
Chr.15: 39945095 - 39949393 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191936.1 NP_001178865.1 1 - 2 335 62

View Full Product Details