Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr93
Entrez Gene ID
499189
Gene Name
WD repeat domain 93
Gene Aliases
RGD1559862
Chromosome Location
Chr.1: 141478655 - 141528160 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191949.1 NP_001178878.1 11 - 12 1245 96
XM_006229401.2 XP_006229463.1 12 - 13 1436 96
XM_006229403.2 XP_006229465.1 12 - 13 1381 96
XM_006229404.2 XP_006229466.1 12 - 13 1436 96
XM_008759547.1 XP_008757769.1 12 - 13 3236 96
XM_008759548.1 XP_008757770.1 11 - 12 1312 96
XM_008759549.1 XP_008757771.1 12 - 13 1385 96

View Full Product Details