Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Aknad1
- Entrez Gene ID
-
691416
- Gene Name
- AKNA domain containing 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 211541828 - 211585332 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073134.1 |
|
7 - 8 |
1604 |
63 |
GenBank mRNA |
BC079051.1 |
- |
7 - 8 |
1604 |
63 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
G-protein signaling modulator 2 |
Gpsm2 |
RGD1560967 |
-36.4kb
|
Anti-sense |
Chr. - 211480120 - 211528105 on Build Rnor_6.0 |
Downstream |
syntaxin binding protein 3 |
Stxbp3 |
Munc18-c |
23.3kb
|
Anti-sense |
Chr. - 211595763 - 211638756 on Build Rnor_6.0 |
View Full Product Details