Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnf4a
Entrez Gene ID
25735
Gene Name
hepatocyte nuclear factor 4, alpha
Gene Aliases
Hnf4, Hnf4alpha
Chromosome Location
Chr.3: 159902441 - 159965003 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270931.1 NP_001257860.1 8 - 9 1067 77
NM_001270933.1 NP_001257862.1 8 - 9 1067 77
NM_022180.2 NP_071516.2 7 - 8 1027 77
XM_006235515.2 XP_006235577.1 7 - 8 1022 77
XM_006235516.2 XP_006235578.1 7 - 8 1022 77
GenBank mRNA D10554.1 - 7 - 8 903 77
EF193390.1 - 7 - 8 848 77
EF193392.1 - 7 - 8 829 77
FJ608816.1 - 8 - 9 1067 77
FJ608817.1 - 8 - 9 1067 77
FJ608818.1 - 8 - 9 1067 77
X57133.1 - 7 - 8 918 77

View Full Product Details