Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abhd11
Entrez Gene ID
360831
Gene Name
abhydrolase domain containing 11
Gene Aliases
Abhd11l1, Wbscr21
Chromosome Location
Chr.12: 24722277 - 24724997 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271180.1 NP_001258109.1 4 - 5 650 67
GenBank mRNA BC088239.1 - 3 - 4 775 67
FQ213935.1 - 4 - 5 641 67

View Full Product Details