Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spock1
Entrez Gene ID
306759
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1
Gene Aliases
-
Chromosome Location
Chr.17: 7675531 - 7797863 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271297.1 NP_001258226.1 - 631 78
GenBank mRNA CB713141.1 - - 326 78

View Full Product Details