Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTA1
Entrez Gene ID
851255
Gene Name
Gene Aliases
FUN39, YAL043C
Chromosome Location
Chr.I: 58695 - 61052 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178188.1 NP_009356.1 1 - 1 2146 88

View Full Product Details