Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEK2
Entrez Gene ID
852248
Gene Name
Gene Aliases
KHD1, YBL032W
Chromosome Location
Chr.II: 160184 - 161329 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178272.1 NP_009521.1 1 - 1 1055 63

View Full Product Details