Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FZO1
Entrez Gene ID
852477
Gene Name
Gene Aliases
YBR179C
Chromosome Location
Chr.II: 586547 - 589114 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178527.1 NP_009738.1 1 - 1 2351 110

View Full Product Details