Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHK1
Entrez Gene ID
852577
Gene Name
Gene Aliases
YBR274W
Chromosome Location
Chr.II: 749594 - 751177 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178622.1 NP_009833.1 1 - 1 1145 68

View Full Product Details