Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GBP2
Entrez Gene ID
850346
Gene Name
Gene Aliases
RLF6, YCL011C
Chromosome Location
Chr.III: 102075 - 103358 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178660.1 NP_009916.1 1 - 1 1150 63

View Full Product Details