Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRE1
Entrez Gene ID
850306
Gene Name
Gene Aliases
YCL051W
Chromosome Location
Chr.III: 35865 - 37616 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178696.1 NP_009879.2 1 - 1 1648 61

View Full Product Details