Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YCP4
Entrez Gene ID
850360
Gene Name
Gene Aliases
YCR004C
Chromosome Location
Chr.III: 119575 - 120318 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178717.1 NP_009930.1 1 - 1 654 67

View Full Product Details