Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC1
Entrez Gene ID
851508
Gene Name
Gene Aliases
YDL052C
Chromosome Location
Chr.IV: 362672 - 363583 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180111.1 NP_010231.1 1 - 1 845 94

View Full Product Details