Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMT1
Entrez Gene ID
851462
Gene Name
Gene Aliases
YDL095W
Chromosome Location
Chr.IV: 287059 - 289512 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180154.1 NP_010188.1 1 - 1 2251 91

View Full Product Details