Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGX2
Entrez Gene ID
851385
Gene Name
Gene Aliases
YDL169C
Chromosome Location
Chr.IV: 158064 - 158735 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180229.1 NP_010112.1 1 - 1 543 86

View Full Product Details