Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAR32
Entrez Gene ID
851381
Gene Name
Gene Aliases
AMU1, YDL173W
Chromosome Location
Chr.IV: 148191 - 149078 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180233.1 NP_010108.1 1 - 1 657 64

View Full Product Details