Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCL9
Entrez Gene ID
851375
Gene Name
Gene Aliases
YDL179W
Chromosome Location
Chr.IV: 138291 - 139205 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180239.1 NP_010102.1 1 - 1 848 80

View Full Product Details