Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMP45
Entrez Gene ID
851304
Gene Name
Gene Aliases
YDL222C
Chromosome Location
Chr.IV: 60872 - 61801 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180282.1 NP_010059.1 1 - 1 854 75

View Full Product Details