Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCS1
Entrez Gene ID
851580
Gene Name
Gene Aliases
YDR017C
Chromosome Location
Chr.IV: 479115 - 482267 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180325.3 NP_010300.3 1 - 1 2949 79

View Full Product Details