Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRM1
Entrez Gene ID
851698
Gene Name
Gene Aliases
YDR120C
Chromosome Location
Chr.IV: 691549 - 693261 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180428.3 NP_010405.3 1 - 1 1654 88

View Full Product Details