Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EKI1
Entrez Gene ID
851725
Gene Name
Gene Aliases
YDR147W
Chromosome Location
Chr.IV: 751631 - 753235 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180454.3 NP_010431.3 1 - 1 1453 113

View Full Product Details