Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARG82
Entrez Gene ID
851753
Gene Name
Gene Aliases
ARGR3, IPK2, YDR173C
Chromosome Location
Chr.IV: 810565 - 811632 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180480.3 NP_010458.3 1 - 1 951 84

View Full Product Details