Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD9
Entrez Gene ID
851803
Gene Name
Gene Aliases
YDR217C
Chromosome Location
Chr.IV: 899551 - 903480 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180525.1 NP_010503.1 1 - 1 3249 75

View Full Product Details