Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD34
Entrez Gene ID
851909
Gene Name
Gene Aliases
YDR314C
Chromosome Location
Chr.IV: 1090433 - 1092511 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180622.1 NP_010600.1 1 - 1 1948 102

View Full Product Details