Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YSP2
Entrez Gene ID
851926
Gene Name
Gene Aliases
LAM2, LTC4, YDR326C
Chromosome Location
Chr.IV: 1120609 - 1124925 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180634.1 NP_010613.1 1 - 1 3549 66

View Full Product Details