Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTP5
Entrez Gene ID
852007
Gene Name
Gene Aliases
YDR398W
Chromosome Location
Chr.IV: 1267471 - 1269402 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180706.1 NP_010686.1 1 - 1 1848 77

View Full Product Details