Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLD5
Entrez Gene ID
852100
Gene Name
Gene Aliases
CDC105, YDR489W
Chromosome Location
Chr.IV: 1431012 - 1431896 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180797.3 NP_010777.3 1 - 1 751 59

View Full Product Details