Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IZH1
Entrez Gene ID
852102
Gene Name
Gene Aliases
YDR492W
Chromosome Location
Chr.IV: 1434924 - 1435874 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180800.1 NP_010780.1 1 - 1 849 75

View Full Product Details