Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVT2
Entrez Gene ID
856645
Gene Name
Gene Aliases
YEL064C
Chromosome Location
Chr.V: 29797 - 31239 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178879.1 NP_010850.1 1 - 1 1249 109

View Full Product Details