Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMI40
Entrez Gene ID
856720
Gene Name
Gene Aliases
YER003C
Chromosome Location
Chr.V: 157736 - 159118 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178894.1 NP_010918.1 1 - 2 31 63

View Full Product Details