Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOT6
Entrez Gene ID
856822
Gene Name
Gene Aliases
PBF2, YER088C
Chromosome Location
Chr.V: 333176 - 335188 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178979.3 NP_011012.3 1 - 1 1744 98

View Full Product Details