Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDA1
Entrez Gene ID
856925
Gene Name
Gene Aliases
YER178W
Chromosome Location
Chr.V: 546817 - 548079 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179068.3 NP_011105.4 1 - 1 755 70

View Full Product Details