Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALR2
Entrez Gene ID
850494
Gene Name
Gene Aliases
YFL050C
Chromosome Location
Chr.VI: 33272 - 35848 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179917.1 NP_116604.1 1 - 1 1947 94

View Full Product Details