Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC1
Entrez Gene ID
850551
Gene Name
Gene Aliases
YFR001W
Chromosome Location
Chr.VI: 149110 - 149724 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179966.1 NP_116656.1 1 - 1 553 91

View Full Product Details