Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MST27
Entrez Gene ID
852830
Gene Name
Gene Aliases
YGL051W
Chromosome Location
Chr.VII: 403687 - 404391 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180916.1 NP_011464.1 1 - 1 242 101

View Full Product Details