Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIF1
Entrez Gene ID
852790
Gene Name
Gene Aliases
YGL090W
Chromosome Location
Chr.VII: 343319 - 344584 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180955.1 NP_011425.1 1 - 1 1147 69

View Full Product Details