Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOS8
Entrez Gene ID
852783
Gene Name
Gene Aliases
YGL096W
Chromosome Location
Chr.VII: 325331 - 326161 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180961.1 NP_011419.1 1 - 1 649 72

View Full Product Details