Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCS3
Entrez Gene ID
852751
Gene Name
Gene Aliases
FIT2B, YGL126W
Chromosome Location
Chr.VII: 270997 - 272139 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180991.3 NP_011389.3 1 - 1 1051 74

View Full Product Details