Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC55
Entrez Gene ID
852685
Gene Name
Gene Aliases
TMR4, YGL190C
Chromosome Location
Chr.VII: 145809 - 147389 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181055.1 NP_011325.1 1 - 1 1452 79

View Full Product Details