Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MDS3
Entrez Gene ID
852678
Gene Name
Gene Aliases
YGL197W
Chromosome Location
Chr.VII: 124698 - 129161 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181062.2 NP_011318.2 1 - 1 4246 71

View Full Product Details