Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLG1
Entrez Gene ID
852657
Gene Name
Gene Aliases
YGL215W
Chromosome Location
Chr.VII: 87981 - 89339 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181080.1 NP_011300.1 1 - 1 752 96

View Full Product Details