Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NQM1
Entrez Gene ID
852934
Gene Name
Gene Aliases
YGR043C
Chromosome Location
Chr.VII: 580435 - 581436 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181172.1 NP_011557.1 1 - 1 944 88

View Full Product Details