Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBF2
Entrez Gene ID
852984
Gene Name
Gene Aliases
YGR092W
Chromosome Location
Chr.VII: 668189 - 669907 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181221.3 NP_011606.3 1 - 1 1648 75

View Full Product Details