Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP7
Entrez Gene ID
852995
Gene Name
Gene Aliases
YGR103W, YPH1
Chromosome Location
Chr.VII: 695417 - 697234 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181232.1 NP_011617.1 1 - 1 1352 104

View Full Product Details