Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HGH1
Entrez Gene ID
853099
Gene Name
Gene Aliases
YGR187C
Chromosome Location
Chr.VII: 870232 - 871416 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181316.3 NP_011703.3 1 - 1 949 106

View Full Product Details