Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLH1
Entrez Gene ID
853187
Gene Name
Gene Aliases
RQT2, YGR271W
Chromosome Location
Chr.VII: 1031791 - 1037694 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181400.4 NP_011787.4 1 - 1 5554 72

View Full Product Details