Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIO2
Entrez Gene ID
853203
Gene Name
Gene Aliases
YGR286C
Chromosome Location
Chr.VII: 1063813 - 1064940 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181415.1 NP_011802.1 1 - 1 1054 66

View Full Product Details