Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VMA10
Entrez Gene ID
856435
Gene Name
Gene Aliases
YHR039C-A, YHR039C-B
Chromosome Location
Chr.VIII: 187173 - 187679 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180027.1 NP_011905.1 1 - 1 253 96

View Full Product Details