Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OSH3
Entrez Gene ID
856472
Gene Name
Gene Aliases
YHR073W
Chromosome Location
Chr.VIII: 242582 - 245572 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179203.1 NP_011940.1 1 - 1 2845 66

View Full Product Details